Learn More
Bulldog Bio Inc Chip Disposable Hemocytometer, NI, 50 slides, 100 Tests
SDP
Catalog No. NC0435502
Encompass_Preferred
C-Chip Disposable Hemocytometer, Neubauer Improved, 100 Tests, for cell counting
Catalog No. NC0435502 Supplier Bulldog Bio Inc Supplier No. DHCN01
C-Chip Disposable Hemocytometer, Neubauer Improved, 100 Tests, for cell counting