Learn More
Grainger Wilton C-Clamp, Forged Steel, 2 in. to 10-1/8 in. Opening, 1 ea.
Catalog No. 502786158
Encompass_Preferred
This extra heavy-duty C-clamp is engineered for demanding industrial environments, including steel fabrication, maintenance, and repair operations. It is constructed from forged steel for exceptional strength and durability during high-pressure clamping tasks.
  • Clamping force of 8,000 lb.
  • Opening capacity of 2 in. to 10-1/8 in.
  • Throat depth of 5-7/8 in.
  • ACME rolled spindle features a protective black oxide coating
  • Replaceable Perma-Pads® with a v-grooved anvil for secure material holding
  • Features a sliding T-handle for ease of use
  • Swivel pad design
Catalog No. 50-278-6158 Supplier Grainger Supplier No. 18G717
This extra heavy-duty C-clamp is engineered for demanding industrial environments, including steel fabrication, maintenance, and repair operations. It is constructed from forged steel for exceptional strength and durability during high-pressure clamping tasks.
  • Clamping force of 8,000 lb.
  • Opening capacity of 2 in. to 10-1/8 in.
  • Throat depth of 5-7/8 in.
  • ACME rolled spindle features a protective black oxide coating
  • Replaceable Perma-Pads® with a v-grooved anvil for secure material holding
  • Features a sliding T-handle for ease of use
  • Swivel pad design