Learn More
Grainger Wilton 100 Series C-Clamp, Forged Steel, 6 to 10 in. Capacity, 1 ea
Catalog No. 502786120
Encompass_Preferred
These heavy-duty C-clamps are designed for rigorous construction and industrial applications. Engineered with a durable drop-forged steel frame and a black oxide finish, they provide high-strength performance and resistance to wear. Each clamp features a precision-machined spindle for reliable operation under demanding conditions.
  • Forged steel frame for heavy-duty structural integrity
  • Black oxide coating provides corrosion resistance
  • Clamping force of 13,750 lb
  • Full-length spindles machined from tempered and hardened cold-drawn steel
  • Square spindle heads designed for heavy wrench tightening
  • Standard black oxide coated swivel perma-pads to minimize workpiece wear
  • Sliding T-handle style for efficient adjustment
Catalog No. 50-278-6120 Supplier Grainger Supplier No. 18G691
These heavy-duty C-clamps are designed for rigorous construction and industrial applications. Engineered with a durable drop-forged steel frame and a black oxide finish, they provide high-strength performance and resistance to wear. Each clamp features a precision-machined spindle for reliable operation under demanding conditions.
  • Forged steel frame for heavy-duty structural integrity
  • Black oxide coating provides corrosion resistance
  • Clamping force of 13,750 lb
  • Full-length spindles machined from tempered and hardened cold-drawn steel
  • Square spindle heads designed for heavy wrench tightening
  • Standard black oxide coated swivel perma-pads to minimize workpiece wear
  • Sliding T-handle style for efficient adjustment