Learn More
Grainger Wilton 800 Series Drop Forged C-Clamp, 1-1/2 to 10 in. Opening, 3-3/4 in. Throat, 1 ea.
Catalog No. 502785696
Encompass_Preferred
This standard depth C-clamp is a drop forged, regular duty tool designed for secure workholding applications. Constructed from high-strength forged steel, it offers reliable performance in environments where robust clamping force is required.
  • Features a 1 1/2 in. to 10 in. jaw opening capacity
  • Includes a 3 3/4 in. throat depth for versatile positioning
  • Delivers 6,000 lb. of clamping force
  • Equipped with replaceable, V-grooved swivel pads
  • Includes black oxide coated steel spindles for increased durability
  • Features a sliding T-handle for ease of use
  • Backed by a lifetime warranty
Catalog No. 50-278-5696 Supplier Grainger Supplier No. 18G709
This standard depth C-clamp is a drop forged, regular duty tool designed for secure workholding applications. Constructed from high-strength forged steel, it offers reliable performance in environments where robust clamping force is required.
  • Features a 1 1/2 in. to 10 in. jaw opening capacity
  • Includes a 3 3/4 in. throat depth for versatile positioning
  • Delivers 6,000 lb. of clamping force
  • Equipped with replaceable, V-grooved swivel pads
  • Includes black oxide coated steel spindles for increased durability
  • Features a sliding T-handle for ease of use
  • Backed by a lifetime warranty