Learn More
Grainger Wilton C-Clamp, Cast Iron, 2-1/2 in. Opening, 1 ea
Catalog No. 502786049
Encompass_Preferred
540A Series C-Clamps are designed for light industrial, woodworking, and repair applications, offering reliable performance for holding objects securely in place. Constructed from durable cast iron, these clamps feature a protective black oxide finish to resist corrosion and ensure long-lasting utility.
  • Opening capacity of 0 to 2 1/2 in.
  • Throat depth of 1 3/4 in.
  • Clamping force of 1,110 lb.
  • Oscillating head with integrated swivel pad for even pressure distribution
  • Sliding T-handle design for efficient adjustment and clamping
  • Durable cast iron body with black oxide finish
Catalog No. 50-278-6049 Supplier Grainger Supplier No. 18G735
540A Series C-Clamps are designed for light industrial, woodworking, and repair applications, offering reliable performance for holding objects securely in place. Constructed from durable cast iron, these clamps feature a protective black oxide finish to resist corrosion and ensure long-lasting utility.
  • Opening capacity of 0 to 2 1/2 in.
  • Throat depth of 1 3/4 in.
  • Clamping force of 1,110 lb.
  • Oscillating head with integrated swivel pad for even pressure distribution
  • Sliding T-handle design for efficient adjustment and clamping
  • Durable cast iron body with black oxide finish