Learn More
Grainger Wilton 540A Series C-Clamp, Cast Iron, 5 in. Opening, 1 ea.
Catalog No. 502787670
Encompass_Preferred
The 540A Series carriage C-clamp is engineered for light industrial, woodworking, and repair tasks. Constructed from durable cast iron, this clamp features a 0 to 5 in. opening capacity and a 2-1/2 in. throat depth to accommodate diverse workpiece sizes.
  • Features a black oxide coating for enhanced durability
  • Equipped with an oscillating head to secure uneven surfaces
  • Includes a sliding cross pin handle for efficient adjustment
  • Provides a clamping force of 2,250 lb.
  • Cast iron frame construction
  • Lifetime warranty
Catalog No. 50-278-7670 Supplier Grainger Supplier No. 18G738
The 540A Series carriage C-clamp is engineered for light industrial, woodworking, and repair tasks. Constructed from durable cast iron, this clamp features a 0 to 5 in. opening capacity and a 2-1/2 in. throat depth to accommodate diverse workpiece sizes.
  • Features a black oxide coating for enhanced durability
  • Equipped with an oscillating head to secure uneven surfaces
  • Includes a sliding cross pin handle for efficient adjustment
  • Provides a clamping force of 2,250 lb.
  • Cast iron frame construction
  • Lifetime warranty