Learn More
Grainger Wilton 540A Series C-Clamp, Cast Iron, 8 in. Opening, 1 ea.
Catalog No. 502786937
Encompass_Preferred
These 540A Series carriage C-clamps are designed for light industrial, woodworking, and repair tasks. Constructed from durable iron, they provide reliable clamping pressure for a variety of projects.
  • Opening capacity of 0 in. to 8 in.
  • Throat depth of 3-1/4 in.
  • Clamping force of 2,750 lb.
  • Black oxide coating for finish protection
  • Oscillating swivel head for secure contact
  • Sliding cross pin handle for ease of adjustment
  • Heavy duty construction
Catalog No. 50-278-6937 Supplier Grainger Supplier No. 18G740
These 540A Series carriage C-clamps are designed for light industrial, woodworking, and repair tasks. Constructed from durable iron, they provide reliable clamping pressure for a variety of projects.
  • Opening capacity of 0 in. to 8 in.
  • Throat depth of 3-1/4 in.
  • Clamping force of 2,750 lb.
  • Black oxide coating for finish protection
  • Oscillating swivel head for secure contact
  • Sliding cross pin handle for ease of adjustment
  • Heavy duty construction