Learn More
Medchemexpress LLC c-di-AMP diammonium | 54447-84-6 | 99.4% | 692.47 | 500 UG
SDP
Catalog No. 5000437961
Encompass_Preferred
c-di-AMP diammonium acts as a STING agonist, stimulating the TBK3-IRF3 signaling pathway to induce the production of type I IFN and TNF. This compound also functions as a crucial bacterial second messenger, influencing cell growth, survival, and virulence, particularly in Gram-positive bacteria, and modulating the host immune response. It is recognized as a potent mucosal adjuvant, capable of eliciting both humoral and cellular immune responses. This product is intended for research use only.
  • Activates TBK3-IRF3 signaling pathway.
  • Induces type I IFN and TNF production.
  • Regulates bacterial cell growth, survival, and virulence.
  • Modulates host immune response.
  • Functions as a potent mucosal adjuvant.
  • Stimulates humoral and cellular immune responses.
Catalog No. 50-004-37961 Supplier Medchemexpress LLC Supplier No. HY12326B500UG
Add to Cart
Add to Cart
c-di-AMP diammonium acts as a STING agonist, stimulating the TBK3-IRF3 signaling pathway to induce the production of type I IFN and TNF. This compound also functions as a crucial bacterial second messenger, influencing cell growth, survival, and virulence, particularly in Gram-positive bacteria, and modulating the host immune response. It is recognized as a potent mucosal adjuvant, capable of eliciting both humoral and cellular immune responses. This product is intended for research use only.
  • Activates TBK3-IRF3 signaling pathway.
  • Induces type I IFN and TNF production.
  • Regulates bacterial cell growth, survival, and virulence.
  • Modulates host immune response.
  • Functions as a potent mucosal adjuvant.
  • Stimulates humoral and cellular immune responses.