Learn More
Medchemexpress LLC C-di-AMP (diammonium) | 99.36% | 692.47 | 50 MG
SDP
Catalog No. 5000436279
Encompass_Preferred
c-di-AMP diammonium is a STING agonist that activates the TBK3-IRF3 signaling pathway, leading to the production of type I IFN and TNF. It also acts as a bacterial second messenger, regulating cell growth, survival, and virulence in Gram-positive bacteria, and modulating the host immune response. This compound functions as a potent mucosal adjuvant, stimulating both humoral and cellular responses.
  • STING agonist
  • Bacterial second messenger regulating cell growth, survival, and virulence in Gram-positive bacteria
  • Regulates host immune response
  • Potent mucosal adjuvant stimulating humoral and cellular responses
Catalog No. 50-004-36279 Supplier Medchemexpress LLC Supplier No. HY12326B50MG
c-di-AMP diammonium is a STING agonist that activates the TBK3-IRF3 signaling pathway, leading to the production of type I IFN and TNF. It also acts as a bacterial second messenger, regulating cell growth, survival, and virulence in Gram-positive bacteria, and modulating the host immune response. This compound functions as a potent mucosal adjuvant, stimulating both humoral and cellular responses.
  • STING agonist
  • Bacterial second messenger regulating cell growth, survival, and virulence in Gram-positive bacteria
  • Regulates host immune response
  • Potent mucosal adjuvant stimulating humoral and cellular responses