Learn More
Medchemexpress LLC c-di-AMP diammonium | 99.4% | 692.47 | 10 MG
SDP
Catalog No. 5000438631
Encompass_Preferred
c-di-AMP diammonium is a STING agonist that activates the TBK3-IRF3 signaling pathway, triggering the production of type I IFN and TNF. It also acts as a bacterial second messenger, regulating cell growth, survival, and virulence in Gram-positive bacteria and influencing host immune response. Furthermore, it serves as a potent mucosal adjuvant, stimulating both humoral and cellular responses.
  • Activates the TBK3-IRF3 signaling pathway.
  • Triggers the production of type I IFN and TNF.
  • Regulates cell growth, survival, and virulence in Gram-positive bacteria.
  • Influences host immune response.
  • Potent mucosal adjuvant, stimulating both humoral and cellular responses.
Catalog No. 50-004-38631 Supplier Medchemexpress LLC Supplier No. HY12326B10MG
c-di-AMP diammonium is a STING agonist that activates the TBK3-IRF3 signaling pathway, triggering the production of type I IFN and TNF. It also acts as a bacterial second messenger, regulating cell growth, survival, and virulence in Gram-positive bacteria and influencing host immune response. Furthermore, it serves as a potent mucosal adjuvant, stimulating both humoral and cellular responses.
  • Activates the TBK3-IRF3 signaling pathway.
  • Triggers the production of type I IFN and TNF.
  • Regulates cell growth, survival, and virulence in Gram-positive bacteria.
  • Influences host immune response.
  • Potent mucosal adjuvant, stimulating both humoral and cellular responses.