Learn More
Medchemexpress LLC c-di-AMP disodium | 2734909-87-4 | 99.14% | 702.38 | 1 MG
SDP
Catalog No. 5000365457
Encompass_Preferred
c-di-AMP (Cyclic diadenylate) sodium is a STING agonist that activates the TBK3-IRF3 signaling pathway, subsequently triggering the production of type I IFN and TNF. It also functions as a bacterial second messenger, primarily in Gram-positive bacteria, where it regulates cell growth, survival, and virulence, and modulates the host immune response. Furthermore, it acts as a potent mucosal adjuvant, stimulating both humoral and cellular responses.
  • Activates the TBK3-IRF3 signaling pathway.
  • Triggers the production of type I IFN and TNF.
  • Regulates cell growth, survival, and virulence in Gram-positive bacteria.
  • Modulates the host immune response.
  • Acts as a potent mucosal adjuvant.
  • Stimulates both humoral and cellular responses.
Catalog No. 50-003-65457 Supplier Medchemexpress LLC Supplier No. 1MGHY12326A
c-di-AMP (Cyclic diadenylate) sodium is a STING agonist that activates the TBK3-IRF3 signaling pathway, subsequently triggering the production of type I IFN and TNF. It also functions as a bacterial second messenger, primarily in Gram-positive bacteria, where it regulates cell growth, survival, and virulence, and modulates the host immune response. Furthermore, it acts as a potent mucosal adjuvant, stimulating both humoral and cellular responses.
  • Activates the TBK3-IRF3 signaling pathway.
  • Triggers the production of type I IFN and TNF.
  • Regulates cell growth, survival, and virulence in Gram-positive bacteria.
  • Modulates the host immune response.
  • Acts as a potent mucosal adjuvant.
  • Stimulates both humoral and cellular responses.