Learn More
Sigma Aldrich Fine Chemicals Biosciences c-di-AMP sodium salt | 54447-84-6 | MFCD28167808 | 1μmol of powder (approx. 0.7mg)
Catalog No. NC1426263
Encompass
c-di-AMP sodium salt | Putity: ≥98% (HPLC) | MW: 658.41 (free acid basis) | 54447-84-6 | MFCD28167808 | 1μmol of powder (approx. 0.7mg)
Catalog No. NC1426263 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SML12311UMO
c-di-AMP sodium salt | Putity: ≥98% (HPLC) | MW: 658.41 (free acid basis) | 54447-84-6 | MFCD28167808 | 1μmol of powder (approx. 0.7mg)