Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-di-IMP (disodium) | 99.9% | 704.35 | 500 UG
SDP

Catalog No. 5000352189
Encompass_Preferred

C-di-IMP (disodium) is a cyclic dinucleotide, chemically known as 3'-Inosinic acid, inosinylyl-(3'5')-, cyclic nucleotide (disodium). This product is suitable for use as a laboratory chemical and in the manufacture of substances.

  • High purity of 99.9% (HPLC)
  • Supplied as a white to off-white solid
  • Molecular weight: 704.35
  • Chemical formula: C20H20N8Na2O14P2

Catalog No. 50-003-52189 Supplier Medchemexpress LLC Supplier No. HY137724A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C-di-IMP (disodium) is a cyclic dinucleotide, chemically known as 3'-Inosinic acid, inosinylyl-(3'5')-, cyclic nucleotide (disodium). This product is suitable for use as a laboratory chemical and in the manufacture of substances.

  • High purity of 99.9% (HPLC)
  • Supplied as a white to off-white solid
  • Molecular weight: 704.35
  • Chemical formula: C20H20N8Na2O14P2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.