Learn More
Eagle Biosciences Inc Fecal C. Difficile Toxin AB Qualitative ELISA Assay Kit
SDP
Supplier:  Eagle Biosciences Inc CDT35K01
Encompass
Clostridium difficile is a common pathogen and a major cause of infectious diarrhea in hospitalized patients. While most strains produce both Toxin A and Toxin B, some only produce one or the other. These enterotoxins, which are both proinflamatory and cytotoxic, attack the mucosal lining of the intestines. If not identified and treated in a timely fashion, can result in permanent damage to the colon or colitis. This assay employs antibodies specific to both Toxin A and Toxin B.
Catalog No. 50-187-8304