Learn More
Eagle Biosciences Inc Fecal C. Difficile Toxin B Qualitative ELISA Assay Kit
SDP
Supplier:  Eagle Biosciences Inc DFB35K01
Encompass
Clostridium difficile is a common pathogen and a major cause of infectious diarrhea in hospitalized patients. While most strains produce both Toxin A and Toxin B, some only produce one or the other. These enterotoxins, which are both proinflamatory and cytotoxic, attack the mucosal lining of the intestines. If not identified and treated in a timely fashion, can result in permanent damage to the colon or colitis. This assay employs an antibody specific to Toxin B without cross-reactivity to toxin A.
Catalog No. 50-187-8303