Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECHLAB™ C. DIFF CHEK™ 60

Catalog No. 23900561
Change view
Click to view available options
Detectable Analytes:
C. difficile GDH Antigen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-900-561 C. difficile GDH Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-561 Supplier TECHLAB™ Supplier No. 30392
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enzyme immunoassay for use as a screening test to detect the C. difficile antigen, glutamate dehydrogenase, in human specimens.

Enzyme immunoassay for use as a screening test to detect the C. difficile antigen, glutamate dehydrogenase (GDH), in fecal specimens from persons suspected of having C. difficile disease.

The glutamate dehydrogenase (GDH) of C. difficile is a good antigen marker for the organism in feces because it is produced in high amounts by all strains, toxigenic or non-toxigenic.

  • Results in approximately 60 min
  • Ideal for facilities currently performing tissue culture assays
  • Easy to perform

Specifications

CPT Code 87449
CE Marker CE-IVD
Clia Complexity High
Detectable Analytes C. difficile GDH Antigen
Detection Method Colorimetric
Format 96-well Plate
Sample Type Stool
Sensitivity >98%
Disposable Single-use
For Use With (Application) ELISA
Sterility Non-sterile
Test Time 60 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.