Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ C. difficile Control Panel (Inactivated Swab)
SDP

Catalog No. 23004142 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
12 Swabs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-004-142 12 Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-004-142 Supplier Microbiologics™ Supplier No. 8227
Add to Cart
Add to Cart
This item is not returnable. View return policy

C. difficile Control Panel (Inactivated Swab) contains six positive control swabs, six negative control swabs, and a Certificate of Analysis.

  • Positive Control (six swabs): Clostridioides difficile
  • Negative Control (six swabs): Clostridium sordellii

Specifications

Biosafety Level 1
Content And Storage 2 to 25°C
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) Clinical
Packaging Type Plastic Box
Quantity 12 Swabs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.