Learn More
Alpco Ltd Clostridium difficile GDH ELISA
SDP
Supplier:  Alpco Ltd 86GDHHUE01
Encompass_Preferred
The Clostridium difficile (C. difficile) GDH ELISA is for the qualitative detection of C. difficile-specific glutamate dehydrogenase in human stool samples. For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2284605