Learn More
Enzo Life Sciences C. difficile toxin B from ribotype 027, (native) (2µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2720002
Encompass_Preferred
Potent enterotoxin. Potent enterotoxin. Alternative name: Clostridium difficile toxin B, tcdB, toxB. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains 0.05M HEPES, 0.15M sodium chloride and 5% sucrose. Source: From Clostridium difficile, strain R20291 (Stoke Mandeville) ribotype 027. Long Term Storage: -80°C. Applications: ELISA LINE assay, Cytotoxicity studies, Antibody capture.
Catalog No. 50-201-4084