Learn More
Alpco Ltd Clostridium difficile Toxin A/B ELISA
SDP
Supplier:  Alpco Ltd 86TABHUE01
Encompass
The Clostridium difficile (C. difficile) Toxin A/B ELISA is for the qualitative determination of C. difficile toxins A and B in human stool and from cultures of toxin-producing C. difficile strains that were previously cultured from stool samples. For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2284612