Learn More
Enzo Life Sciences C. difficile toxoid A (50µg)
SDP
Supplier:  Enzo Life Sciences BMLG1450050
Encompass_Preferred
Alternative name: Clostridium difficile toxoid A. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized. Contains 0.05M HEPES, 0.15M sodium chloride and 5% sucrose. Source: From Clostridium difficile, strain VPI10463. Inactivated by formaldehyde. Long Term Storage: +4°C. Applications: ELISA LINE assay, Immunogen, Cytotoxicity studies, Antibody capture.
Catalog No. 50-201-0846