Learn More
Enzo Life Sciences C-erbB-2 (human) clonal antibody (A24-V) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX810227L001
Encompass_Preferred
Clone: A24-V. Isotype: IgG. Immunogen: Synthetic peptide corresponding to the C-terminal region of human C-erbB-2. Species reactivity: Human. Applications: IHC (FS), IHC (PS). Purity: Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. Formulation: Liquid. In 20mM TRIS/HCl, pH 8.0, containing 20mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-0086