Learn More
Enzo Life Sciences c-erbB-2 (human), (recombinant) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4600010
Encompass_Preferred
Alternative name: ErbB2, Her2, Neu, CD340. Source: Produced in insect cells. Active c-erbB2 (aa 679-1255). Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2035