Learn More
Medchemexpress LLC C-erbB-4/HER4 antibody (YA456) | Predicted band size: 147 kDa; Observed band size: 180 kDa | 50 UL
SDP
Catalog No. 5000396058
Encompass_Preferred
C-erbB-4/HER4 Antibody (YA456) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting C-erbB-4/HER4. It is for research use only. The antibody is recombinant and monoclonal.
  • Reacts with human and rat species.
  • Immunogen used is a synthetic peptide corresponding to human ErbB4 aa721-972.
  • Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA with 0.01% Sodium azide as preservative.
  • Should be stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles.
  • Shipping is with blue ice.
  • Validated for applications such as Western Blot (WB) with a dilution ratio of 1:500-1:1000.
Catalog No. 50-003-96058 Supplier Medchemexpress LLC Supplier No. HYP8066050UL
Add to Cart
Add to Cart
C-erbB-4/HER4 Antibody (YA456) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting C-erbB-4/HER4. It is for research use only. The antibody is recombinant and monoclonal.
  • Reacts with human and rat species.
  • Immunogen used is a synthetic peptide corresponding to human ErbB4 aa721-972.
  • Supplied in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA with 0.01% Sodium azide as preservative.
  • Should be stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles.
  • Shipping is with blue ice.
  • Validated for applications such as Western Blot (WB) with a dilution ratio of 1:500-1:1000.