Learn More
Cole-Parmer C-Flex 072 TPE Tubing, 1/8" ID x 1/4" OD, 50 Ft; Saint Gobain 072-125-2
Catalog No. 5000610335 Shop All Cole-Parmer Products
Encompass_Preferred
Excellent heat seal and sterile weld capabilities make it ideal for biopharmaceutical applications
  • Flexible and durable for use in peristaltic pumps
  • Less gas permeable compared to silicone
  • Opaque tubing—perfect for light sensitive fluids
  • USP Class VI and European Pharmacopeia 3.2.9 compliant
  • 1/8" ID x 1/4" OD
  • Easily meet the critical demands of pharmaceutical and biopharmaceutical applications with C-Flex TPE tubing. This moldable and bondable tubing is specifically designed for aseptic welding connections and sealing disconnection; seal or weld before or after sterilization depending on your application requirements.
    Excellent flexibility and durability offer prolonged service life in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability characteristics protects sensitive fluids from gas ingress.
    Catalog No. 50-006-10335 Supplier Cole-Parmer Supplier No. 5012309
    Excellent heat seal and sterile weld capabilities make it ideal for biopharmaceutical applications
  • Flexible and durable for use in peristaltic pumps
  • Less gas permeable compared to silicone
  • Opaque tubing—perfect for light sensitive fluids
  • USP Class VI and European Pharmacopeia 3.2.9 compliant
  • 1/8" ID x 1/4" OD
  • Easily meet the critical demands of pharmaceutical and biopharmaceutical applications with C-Flex TPE tubing. This moldable and bondable tubing is specifically designed for aseptic welding connections and sealing disconnection; seal or weld before or after sterilization depending on your application requirements.
    Excellent flexibility and durability offer prolonged service life in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability characteristics protects sensitive fluids from gas ingress.