Learn More
Accuflow Systems C-Flex™ 082 formulation tubing, 50-foot, ID 0.063"" x OD 0.188"" x Wall 0.063"" is manufactured from pharmaceutical-grade thermoplastic elastomer, fully validated and tested to a variety of specifications.
Catalog No. 501539359
Encompass_Preferred
C-Flex is the original patented thermoplastic elastomer tubing specifically designed to meet the critical demands of pharmaceutical and biopharmaceutical applications for fluid processing. C-Flex tubing for Biopharma Manufacturing is specifically designed for aseptic welding connections and sealing disconnection. C-Flex is suitable for sterilization by gamma irradiation and autoclave. C-Flex is moldable, bondable and formable ideal for single-use assemblies, overmolds, tubing and bags manifolds. C-Flex tubing is manufactured from pharmaceutical-grade thermoplastic elastomer, is fully characterized, validated and tested to a variety of specifications. Extensive biological, chemical, physical properties and extractable testing is available to assist in the validation of C-Flex application. Sealable and weldable either pre- or post-sterilization. Sterilizable by gamma irradiation and autoclave. Complies with USP Class VI and ISO standards.
Catalog No. 50-153-9359 Supplier Accuflow Systems Supplier No. 0820632
C-Flex is the original patented thermoplastic elastomer tubing specifically designed to meet the critical demands of pharmaceutical and biopharmaceutical applications for fluid processing. C-Flex tubing for Biopharma Manufacturing is specifically designed for aseptic welding connections and sealing disconnection. C-Flex is suitable for sterilization by gamma irradiation and autoclave. C-Flex is moldable, bondable and formable ideal for single-use assemblies, overmolds, tubing and bags manifolds. C-Flex tubing is manufactured from pharmaceutical-grade thermoplastic elastomer, is fully characterized, validated and tested to a variety of specifications. Extensive biological, chemical, physical properties and extractable testing is available to assist in the validation of C-Flex application. Sealable and weldable either pre- or post-sterilization. Sterilizable by gamma irradiation and autoclave. Complies with USP Class VI and ISO standards.