Learn More
Chemglass Life Sciences C-Flex, 36" Extension Line, Plugged Ends, Sterile
SDP
Catalog No. 5000083111
Encompass
Fluid Transfer Extension Lines are produced in a Class 7 Cleanroom with weldable size 16 (1/8 x 1/4) TPE tubing. They are available in both Advantaflexand C-Flex tubing, both with plugged ends. The Fluid transfer lines are designed to aid in the process of transfer and filling of bioreactors, mixers and fermenters. Individually wrapped and sterile.
Catalog No. 50-000-83111 Supplier Chemglass Life Sciences Supplier No. CLS1390C036
Fluid Transfer Extension Lines are produced in a Class 7 Cleanroom with weldable size 16 (1/8 x 1/4) TPE tubing. They are available in both Advantaflexand C-Flex tubing, both with plugged ends. The Fluid transfer lines are designed to aid in the process of transfer and filling of bioreactors, mixers and fermenters. Individually wrapped and sterile.