Learn More
Cole-Parmer C-Flex 374 TPE Tubing, 1/16" ID x 3/16" OD, 50 Ft; Saint Gobain 374-063-2
Catalog No. 5000610271 Shop All Cole-Parmer Products
Encompass_Preferred
Excellent heat seal and sterile weld capabilities make it ideal for biopharma applications
  • Flexible and durable for use in peristaltic pumps
  • Less gas permeable compared to silicone
  • Translucent tubing for flow visibility
  • UPS Class VI and European Pharmacopeia 3.2.9 compliant
  • 1/16" ID x 3/16" OD
  • Easily meet the critical demands of pharmaceutical and biopharma applications with C-Flex TPE tubing. This moldable and bondable tubing is specifically designed for aseptic welding connections and sealing disconnection; seal or weld before or after sterilization depending on your application requirements.
    Excellent flexibility and durability offer prolonged service life in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability characteristics protects sensitive fluids from gas ingress. Translucent tubing enables visual monitoring of the flow, so you can control or adjust the flow during the process.
    Catalog No. 50-006-10271 Supplier Cole-Parmer Supplier No. 5012329
    Excellent heat seal and sterile weld capabilities make it ideal for biopharma applications
  • Flexible and durable for use in peristaltic pumps
  • Less gas permeable compared to silicone
  • Translucent tubing for flow visibility
  • UPS Class VI and European Pharmacopeia 3.2.9 compliant
  • 1/16" ID x 3/16" OD
  • Easily meet the critical demands of pharmaceutical and biopharma applications with C-Flex TPE tubing. This moldable and bondable tubing is specifically designed for aseptic welding connections and sealing disconnection; seal or weld before or after sterilization depending on your application requirements.
    Excellent flexibility and durability offer prolonged service life in peristaltic pumps and help minimize downtime due to tubing failures. The low permeability characteristics protects sensitive fluids from gas ingress. Translucent tubing enables visual monitoring of the flow, so you can control or adjust the flow during the process.