Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Single-Use Transfer Tubing, Gamma-Irradiated C-Flex, 1/8"" ID x 1/4"" OD; 25 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX9612405
Encompass
Masterflex gamma-irradiated transfer tubing protects your sensitive processes and products from accidental contamination while saving time! Gamma-irradiated tubing helps you avoid costly delays that can be associated with outsourced gamma irradiation services. Tubing is double bagged prior to undergoing the gamma irradiation sterilization process, and all sealed tubing is gamma irradiated at 32.5 kGy to offer optimal microbial control in a critical fluid pathway. Tubing boxes include gamma radiation-sensitive labels to confirm treatment. Ideal for pharmaceutical and biopharmaceutical processing, sensitive research, single-use systems, and more. Masterflex C-Flex tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic ¿ protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production.
Catalog No. NC1496601