Learn More
AVANTOR MASTERFLEX (VWR LLC) C-Flex Animal-Derived Component Free (ADCF) 1/8 x 1/4"" Tubing, 50Ft
Catalog No. NC0381278
Encompass
Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent. Certification: Resin meets USP and Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC0381278 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643250
Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent. Certification: Resin meets USP and Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: Autoclave, ethylene oxide, or gamma irradiation.