Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, C-Flex, Opaque White, 1/2"" ID x 3/4""OD; 25 ft
Catalog No. NC9477543
Encompass
Applications: General-purpose lab uses requiring high performance and long life. Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Opaque white. Certification: Resin meets USP Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: With autoclave, ethylene oxide, or gamma irradiation.
Catalog No. NC9477543 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0642481
Applications: General-purpose lab uses requiring high performance and long life. Characteristics: Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Opaque white. Certification: Resin meets USP Class VI requirements. Tubing is manufactured according to GMP. Temperature range: -50 to 275¿F (-45 to 135¿C) Sterilize: With autoclave, ethylene oxide, or gamma irradiation.