Learn More
Medchemexpress LLC HY-18791 5mg Medchemexpress, c-Fms-IN-1 CAS:885703-64-0 Purity:>98%
SDP
Catalog No. 501932823
Encompass_Preferred
Medchemexpress, HY-18791 5mg c-Fms-IN-1 CAS:885703-64-0 c-Fms-IN-1 is a FMS kinase inhibitor with an IC 50 of 0.0008 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2823 Supplier Medchemexpress LLC Supplier No. HY187915MG
Medchemexpress, HY-18791 5mg c-Fms-IN-1 CAS:885703-64-0 c-Fms-IN-1 is a FMS kinase inhibitor with an IC 50 of 0.0008 μM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.