Learn More
Medchemexpress LLC c-Fms-IN-10 | 1527517-50-5 | 98.6% | 429.5 g/mol | C22H19N7OS | 50MG
SDP
Catalog No. 5000236895
Encompass_Preferred
c-Fms-IN-10 is a potent small-molecule inhibitor of the Colony Stimulating Factor 1 Receptor (CSF-1R, also called c-Fms), developed as a thieno[3,2-d]pyrimidine derivative with reported anti-tumor activity and an in vitro IC50 of 2 nM. It is supplied as a solid and as a 10 mM solution in DMSO for in vitro use, and has high purity and well-defined storage recommendations for both powder and solution forms.
  • Potent inhibitor of CSF-1R with IC50 = 2 nM.
  • Thieno[3,2-d]pyrimidine derivative with kinase inhibitory activity.
  • Available as solid solids and as a 10 mM DMSO solution for in vitro studies.
  • Soluble in DMSO at 10 mg/mL using ultrasonic warming to 60°C.
  • High purity suitable for research applications.
  • Defined storage stability for powder and solution forms.
Catalog No. 50-002-36895 Supplier Medchemexpress LLC Supplier No. HY12629750MG
c-Fms-IN-10 is a potent small-molecule inhibitor of the Colony Stimulating Factor 1 Receptor (CSF-1R, also called c-Fms), developed as a thieno[3,2-d]pyrimidine derivative with reported anti-tumor activity and an in vitro IC50 of 2 nM. It is supplied as a solid and as a 10 mM solution in DMSO for in vitro use, and has high purity and well-defined storage recommendations for both powder and solution forms.
  • Potent inhibitor of CSF-1R with IC50 = 2 nM.
  • Thieno[3,2-d]pyrimidine derivative with kinase inhibitory activity.
  • Available as solid solids and as a 10 mM DMSO solution for in vitro studies.
  • Soluble in DMSO at 10 mg/mL using ultrasonic warming to 60°C.
  • High purity suitable for research applications.
  • Defined storage stability for powder and solution forms.