Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C-fms-in-10 | 1527517-50-5 | 98.6% | 429.50 g·mol^-1 | C22H19N7OS | 10MG
SDP

Catalog No. 5000235580
Encompass_Preferred

c-Fms-IN-10 is a thieno[3,2-d]pyrimidine-derived small-molecule inhibitor of the colony-stimulating factor-1 receptor (CSF-1R/c-Fms) that exhibits potent kinase inhibition (IC50 = 2 nM) and reported antitumor activity. The compound is supplied as a powder for biochemical and cellular research, and manufacturer-provided solubility and storage guidance facilitate experimental use.

  • Potent CSF-1R inhibitor with IC50 of 2 nM.
  • Thieno[3,2-d]pyrimidine-derived small molecule.
  • High reported purity of 98.6% for consistent assay performance.
  • Soluble in DMSO at 10 mg/mL with ultrasonic warming.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (2 years).
  • Includes in vivo formulation protocols for suspension preparation.

Catalog No. 50-002-35580 Supplier Medchemexpress LLC Supplier No. HY12629710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

c-Fms-IN-10 is a thieno[3,2-d]pyrimidine-derived small-molecule inhibitor of the colony-stimulating factor-1 receptor (CSF-1R/c-Fms) that exhibits potent kinase inhibition (IC50 = 2 nM) and reported antitumor activity. The compound is supplied as a powder for biochemical and cellular research, and manufacturer-provided solubility and storage guidance facilitate experimental use.

  • Potent CSF-1R inhibitor with IC50 of 2 nM.
  • Thieno[3,2-d]pyrimidine-derived small molecule.
  • High reported purity of 98.6% for consistent assay performance.
  • Soluble in DMSO at 10 mg/mL with ultrasonic warming.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (2 years).
  • Includes in vivo formulation protocols for suspension preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.