Learn More
Medchemexpress LLC HY-18787 1mg Medchemexpress, c-Fms-IN-2 CAS:791587-67-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY187871MG
Encompass_Preferred
Medchemexpress, HY-18787 1mg c-Fms-IN-2 CAS:791587-67-2 c-Fms-IN-2 is a FMS kinase inhibitor with an IC50 of 0.024 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2089
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.