Learn More
TARGETMOL CHEMICALS INC C-FMS-IN-2 5MG
SDP
Catalog No. 502581669
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. c-Fms-IN-2 is an inhibitor of c-FMS kinase (IC50 = 24 nM). purity: 97%
Catalog No. 50-258-1669 Supplier TARGETMOL CHEMICALS INC Supplier No. T107755MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. c-Fms-IN-2 is an inhibitor of c-FMS kinase (IC50 = 24 nM). purity: 97%