Learn More
Cell Signaling Technology c-Fos Mouse Monoclonal Antibody, Unconjugated, Mouse IgG2a, 100 μg
SDP
Catalog No. 503133996
Encompass
This mouse monoclonal antibody is designed to detect endogenous levels of total c-Fos protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of human c-Fos. This carrier-free formulation is devoid of BSA and azide, making it suitable for specialized or custom antibody labeling applications, such as conjugation to fluorophores, metals, lanthanides, or oligonucleotides.
  • Validated for use in ELISA and/or ELISA-like assays
  • Provided in a 1X PBS buffer (pH 7.8)
  • Ideal for technologies requiring custom labeling
  • Tested by size exclusion chromatography to ensure antibody integrity
  • Species reactivity confirmed for human, mouse, and monkey
Catalog No. 50-313-3996 Supplier Cell Signaling Technology Supplier No. 16282SF
This mouse monoclonal antibody is designed to detect endogenous levels of total c-Fos protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues near the amino terminus of human c-Fos. This carrier-free formulation is devoid of BSA and azide, making it suitable for specialized or custom antibody labeling applications, such as conjugation to fluorophores, metals, lanthanides, or oligonucleotides.
  • Validated for use in ELISA and/or ELISA-like assays
  • Provided in a 1X PBS buffer (pH 7.8)
  • Ideal for technologies requiring custom labeling
  • Tested by size exclusion chromatography to ensure antibody integrity
  • Species reactivity confirmed for human, mouse, and monkey