Learn More
Curiox Biosystems Inc C-FREE PLUTO SYSTEM LIQUID HA
SDP
Catalog No. NC3644128
Encompass
  • High-throughput automated liquid handling system
  • C-FREE™ laminar flow technology
  • Centrifuge-free cell washing, staining, and antibody cocktailing
  • Four varispan- and 96-channel pipette heads
  • Throughput: Up to 96 wells
  • Gentle cell handling that preserves viability and integrity
  • Combines liquid handling, antibody cocktail preparation, cell washing
  • Fully automated with pre-installed one-touch protocols
  • Enhanced reproducibility and reduced human error
  • Wash cycle time: 3 to 30 minutes
  • Antibody cocktailing capacity: Up to 96 antibodies
  • Compatible with standard-format 96-well microplates, deep-well plates, Laminar Wash™ plates
  • Compact footprint, replaces bulky hardware
  • Suitable for flow cytometry, mass cytometry, multiplex immunoassays, single-cell sequencing, and more
  • 21CFR Part 11 compliant
Catalog No. NC3644128 Supplier Curiox Biosystems Inc Supplier No. CFREE PLUTO HT SYSTEM
Add to Cart
Add to Cart
Request a Quote
  • High-throughput automated liquid handling system
  • C-FREE™ laminar flow technology
  • Centrifuge-free cell washing, staining, and antibody cocktailing
  • Four varispan- and 96-channel pipette heads
  • Throughput: Up to 96 wells
  • Gentle cell handling that preserves viability and integrity
  • Combines liquid handling, antibody cocktail preparation, cell washing
  • Fully automated with pre-installed one-touch protocols
  • Enhanced reproducibility and reduced human error
  • Wash cycle time: 3 to 30 minutes
  • Antibody cocktailing capacity: Up to 96 antibodies
  • Compatible with standard-format 96-well microplates, deep-well plates, Laminar Wash™ plates
  • Compact footprint, replaces bulky hardware
  • Suitable for flow cytometry, mass cytometry, multiplex immunoassays, single-cell sequencing, and more
  • 21CFR Part 11 compliant