Learn More
AMID BIOSCIENCES Amid Biosciences, C-His-pTrham Rapid Cloning and Gene Expression Kit, 1 Kit
SDP
Catalog No. NC4043484
Encompass_Preferred
C-His-pTrham rapid cloning and gene expression kit is supplied as a 1 Kit format for construction and bacterial expression of proteins with a C-terminal His-tag.The C-His-pTrham Cloning Kit consists of linearized C-His-pTrham Vector that allows cloning of any PCR product, E. coli AB 5-alpha Chemically Competent Cells for cloning and protein expression, control insert DNA, and sequencing primers for clone analysis. Cloning into C-His-pTrham vector results to production of recombinant proteins fused only to a 6X histidine tag sequence at the C-terminus.- Quantity - 1 kit- Main benefits of this kit include a universal ability to clone any insert, no further enzymatic step after PCR amplification, no extra nucleotides, and greater than 95% positive coloniesThis product is for research use only.
Catalog No. NC4043484 Supplier AMID BIOSCIENCES Supplier No. CHPTR401
C-His-pTrham rapid cloning and gene expression kit is supplied as a 1 Kit format for construction and bacterial expression of proteins with a C-terminal His-tag.The C-His-pTrham Cloning Kit consists of linearized C-His-pTrham Vector that allows cloning of any PCR product, E. coli AB 5-alpha Chemically Competent Cells for cloning and protein expression, control insert DNA, and sequencing primers for clone analysis. Cloning into C-His-pTrham vector results to production of recombinant proteins fused only to a 6X histidine tag sequence at the C-terminus.- Quantity - 1 kit- Main benefits of this kit include a universal ability to clone any insert, no further enzymatic step after PCR amplification, no extra nucleotides, and greater than 95% positive coloniesThis product is for research use only.