Learn More
Medchemexpress LLC C.I. Basic red 14 | 12217-48-0 | 99.09% | 25 G
SDP
Catalog No. 5000414208
Encompass_Preferred
C.I. Basic red 14 is an alkaline dye that can be removed from aqueous solutions in intermittent adsorption systems using tree ferns as biosorbents. It is for research use only.
  • Functions as an alkaline dye
  • Can be removed from aqueous solutions using biosorbents
  • Suitable for research applications
  • Categorized as a fluorescent dye
  • Used in labeling chemistry
  • Functions as a chemical stain
  • Utilized as an analysis chemical
  • Belongs to analytical dyes classification
Catalog No. 50-004-14208 Supplier Medchemexpress LLC Supplier No. 25GHYD0478
C.I. Basic red 14 is an alkaline dye that can be removed from aqueous solutions in intermittent adsorption systems using tree ferns as biosorbents. It is for research use only.
  • Functions as an alkaline dye
  • Can be removed from aqueous solutions using biosorbents
  • Suitable for research applications
  • Categorized as a fluorescent dye
  • Used in labeling chemistry
  • Functions as a chemical stain
  • Utilized as an analysis chemical
  • Belongs to analytical dyes classification