Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC C.I. Disperse blue 79 | 12239-34-8 | 100 MG
SDP

Catalog No. 5000409817
Encompass_Preferred

C.I. Disperse blue 79 is a blue dye commonly used for fiber dyeing.

  • For research use only.
  • Appearance: Solid.
  • Color: Brown to black.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light.
  • In solvent storage: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-004-09817 Supplier Medchemexpress LLC Supplier No. HYD0491100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C.I. Disperse blue 79 is a blue dye commonly used for fiber dyeing.

  • For research use only.
  • Appearance: Solid.
  • Color: Brown to black.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, protect from light.
  • In solvent storage: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.