Learn More
Enzo Life Sciences c-IAP1 monoclonal antibody (1E1-1-10) (100 µg)
SDP
Supplier:  Enzo Life Sciences ALX803335C100
Encompass_Preferred
Clone: 1E1-1-10. Host: Rat. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 221-232 of mouse c-IAP1. UniProt: Q62210. Species Reactivity: Human, Mouse. Applications: WB. Recommended dilutions: Western Blot (1:1,000-1:4,000 using ECL; suggested blocking and dilution buffer is PBST containing 0.1% Tween 20 and either 5% skim milk or 5% BSA. Suggested incubation time is 1 hour at room temperature or overnight at 4°C.). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. 0.2μm-filtered solution in PBS containing 0.05% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. NC1413148