Learn More
Enzo Life Sciences c-IAP2 monoclonal antibody (human) (16E-6-3) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX803341C100
Encompass_Preferred
Clone: 16E-6-3. Isotype: IgG1. Immunogen: GST-fusion product of the CARD domain of human c-IAP2. Species reactivity: Human. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. 0.2µm-filtered solution in PBS. .
Catalog No. 50-200-9741