Learn More
Bioworld C-JUN (RECOMBINANT)
Supplier:  Bioworld 220600341
Encompass_Preferred
Recombinant Human C-JUN Source: E.Coli Background: C-JUN is a gene which, in combination with c-Fos, forms the AP-learly response transcription factor. C-JUN is activated by the JNK pathway. C-JUN is the putative transforming gene of...
Catalog No. 50-253-130