Learn More
Eurofins Discoverx c-Kit, active, 10 µg. (14-559)
Supplier:  Eurofins Discoverx 14559
Encompass
N-terminal GST-tagged recombinant, human Kit amino acids 544-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8969