Learn More
Enzo Life Sciences c-Kit polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADIKAPTK005E
Encompass_Preferred
Host: Rabbit. Isotype: IgG. Immunogen: Synthetic peptide for C-terminal of human c-Kit. UniProt ID: P10721. Species reactivity: Human. Applications: IHC (PS), WB. Recommended Dilutions: Immunohistochemistry (paraffin sections, 1:200) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-200-7311