Learn More
Sino Biological Recombinant Human BTK Protein (Full Length, R562P, His Tag)
Catalog No. 502575776
Encompass_Preferred
Recombinant full length human BTK (R562P) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.
Catalog No. 50-257-5776 Supplier Sino Biological Supplier No. B1012DH500
Recombinant full length human BTK (R562P) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag.