Learn More
GA International Cryogenic Tamper-Evident Tape 0.875" x 50' / 22.2mm x 15m White 1" / 25.4mm From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC2310208
Encompass_Preferred
Tamper-evident cryogenic tape for sealing ultra-low temperature freezer and cryogenic containers such as cryo vials, cryo/freezer boxes, cryo cassettes, and tubes. The continuous tape will tear-up upon any attempt to remove it from most surfaces, indicating whether the contents have been compromised. Durable and waterproof, the tamper-evident tape is available in a range of colors for easy identification. Writable using our permanent cryogenic markers, the cryo tape is suitable for long-term storage in liquid nitrogen (-196°C), laboratory freezers (-80°C), and for transportation on dry ice.
Catalog No. NC2310208 Supplier GA International Supplier No. TETA22C150WH
Tamper-evident cryogenic tape for sealing ultra-low temperature freezer and cryogenic containers such as cryo vials, cryo/freezer boxes, cryo cassettes, and tubes. The continuous tape will tear-up upon any attempt to remove it from most surfaces, indicating whether the contents have been compromised. Durable and waterproof, the tamper-evident tape is available in a range of colors for easy identification. Writable using our permanent cryogenic markers, the cryo tape is suitable for long-term storage in liquid nitrogen (-196°C), laboratory freezers (-80°C), and for transportation on dry ice.