Learn More
C.L. Sturkey Diamond Disposable Microtome Blades
Catalog No. 22210011 Shop All CL Sturkey Products
Change view
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
170 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Profile Quantity
22-210011 Low 170 Pk.
22-210014 High 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-210011 Supplier C.L. Sturkey Supplier No. DT315D
Add to Cart
Add to Cart
Unique blade that has a special effectiveness on hard tissue and decalcified bone
Features an amorphous diamond coating that is applied to the cutting edge, resulting in a very tough and durable blade
Packaging Options
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile Low
Disposable Single-use
Quantity 170 Pk.
Coating Amorphous Diamond