Learn More
C.L. Sturkey Diamond Disposable Microtome Blades
Catalog No. 22210014 Shop All CL Sturkey Products
Click to view available options
Profile:
High
Low
Quantity:
100 Pk.
170 Pk.
Catalog No. 22-210014
Supplier: C.L. Sturkey D554D
Only null left
Add to Cart
Add to Cart
Unique blade that has a special effectiveness on hard tissue and decalcified bone
Features an amorphous diamond coating that is applied to the cutting edge, resulting in a very tough and durable blade
Packaging Options
  • Bulk Dispenser: Holds 120 Heavy Duty High-Profile or 200 Low-Profile blades per dispenser
  • Box Packaging: Holds 100 Heavy Duty High-Profile or 170 Low-Profile blades per box (non-dispensing)
Specifications
Profile High
Disposable Single-use
Quantity 100 Pk.
Coating Amorphous Diamond
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.